ContactUsLinkComponent
VWR®, Filling and Venting Cap for VWR® Carboys
VWR®, Filling and Venting Cap for VWR® Carboys
Catalog # 216-1842
Supplier:  VWR Collection
VWR®, Filling and Venting Cap for VWR® Carboys
Catalog # 216-1842
Supplier:  VWR Collection
Specifications
  • Description:
    Filling/venting 3-port cap for VWR® carboys
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
The filling and venting cap for VWR® carboys is an autoclavable closure designed for the aseptic transfer of media, biological reagents, pure water and chemicals
Each cap is equipped with integrated 6,35 mm ports.