ContactUsLinkComponent
J.T.Baker® BAKERBOND®, Wide-Pore Ethyl (C₂) Reversed Phase Media
J.T.Baker® BAKERBOND®, Wide-Pore Ethyl (C₂) Reversed Phase Media
Catalog # 7199-00
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKERBOND®, Wide-Pore Ethyl (C₂) Reversed Phase Media
Catalog # 7199-00
Supplier:  Avantor
CAS Number:  
Specifications
  • Particle Size:
    40 µm
  • Pk:
    100 g
  • Pore size:
    60 Å
  • Description:
    Ethyl (C₂), BAKERBOND®