TKN indicator solution
705-0112
: Hach
:
- Pk:50 mL
- Description:TKN indicator solution
Indicator solution for pH adjustment in total kjeldahl nitrogen determination by the Nessler method.
: Packed in a self-contained dropping bottle.
Indicator solution for pH adjustment in total kjeldahl nitrogen determination by the Nessler method.