Capsule filters, Polycap™ SPF, Whatman™
These Polycap™ SPF capsule filters are optimised for prefiltration applications. They are typically used upstream of a Polycap™ AS or Polycap™ TC capsule during enzyme, nutrient media or buffer filtration.
- Three layers of filter media: fine glass microfiber (GMF), ultrafine GMF, and polyethersulfone (PES)
- Suitable for hard-to-filter solutions, particularly sera and protein solutions
- Can be sterilized by autoclaving
- Well suited for serum, viral suspension, nutrient, biological, immunological, enzyme, and buffer filtration
Biological filtration, especially serum filtration, is often difficult because of the high concentration of complex particulates, lipids, triglycerides, and lipoproteins in solution that can clog filters. Single-layer membrane filters clog rapidly without proper prefiltration.
Polycap™ SPF prefilters are highly suitable for biological filtration, especially serum, buffer, and enzyme filtration, because of the density graded pre-filtration layers they contain. Prefiltration with Polycap™ SPF can help extend the life of the final filter.
: Manufactured according to ISO9001:2008 QMS standards
- Catalog No:
- 515-0107
- 515-0110
- Type:
- Polycap™ SPF 36, PES, with glass fibre prefilter
- Polycap™ SPF 75, PES, with glass fibre prefilter
- Filtration area:
- 260 cm²
- 535 cm²
- Max. pressure:
- 4,1 bar
- 4,1 bar
Frequently Bought Together