Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 1 mol/l (2 N), Titripur®, Supelco®
Sulphuric acid 1 mol/l (2 N), Titripur®, Supelco®
Catalog # 1.60313.1003
Supplier:  Merck
CAS Number:  
Sulphuric acid 1 mol/l (2 N), Titripur®, Supelco®
Catalog # 1.60313.1003
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle with RFID tag
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Form:
    Liquid
  • Concentration:
    0,995 - 1,005 mol/l
  • Measurement uncertainty:
    ±0,003 mol/l
  • Traceability:
    NIST SRM

Specification Test Results

About this item

 :
 :98.08 g/mol
UN:2796
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Boiling Pt:330
Melting Pt:10.38
Density:1.1 g/cm³ (20 °C)
ADR:8
 :M-Clarity™ quality level = MQ100