0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Sodium carbonate
Sodium carbonate solution 0.05 mol/l (0.1 N), Titripur®, Supelco®
Sodium carbonate solution 0.05 mol/l (0.1 N), Titripur®, Supelco®
1.60310.1000
:
Merck
:
Sodium carbonate solution 0.05 mol/l (0.1 N), Titripur®, Supelco®
1.60310.1000
:
Merck
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
105.99 g/mol
:
M-Clarity™ quality level = MQ100
:
Ambient
:
497-19-8
:
Na₂CO₃
:
MFCD00003494