Order Entry
Belgium
ContactUsLinkComponent
Color-WP, certified reference material (CRM), Supelco®
Color-WP, certified reference material (CRM), Supelco®
Catalog # QC1126-20ML
Supplier:  Merck
CAS Number:  
Color-WP, certified reference material (CRM), Supelco®
Catalog # QC1126-20ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    20 mL
  • Description:
    Color-WP

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.