Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 0.100 N volumetric solution
Corrosive
Sulphuric acid 0.100 N volumetric solution
Catalog # HACH20253
Supplier:  Hach
CAS Number:  
undefined
Corrosive
Sulphuric acid 0.100 N volumetric solution
Catalog # HACH20253
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :98.08 g/mol
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589