Order Entry
Belgium
ContactUsLinkComponent
Potassium dihydrogen phosphate concentrate for dilution to pH 7,4, DILUT-IT® dissolution media concentrate, J.T.Baker®
Potassium dihydrogen phosphate concentrate for dilution to pH 7,4, DILUT-IT® dissolution media concentrate, J.T.Baker®
Supplier:  Avantor
Certificates
Potassium dihydrogen phosphate concentrate for dilution to pH 7,4, DILUT-IT® dissolution media concentrate, J.T.Baker®
Supplier:  Avantor

Specification Test Results

  • Assay (M) at 26X Dilution (as Potassium Phosphate Monobasic):
    0.045 - 0.054
  • Appearance:
    Passes Test
  • pH at 25 °C at 26X Dilution:
    7.35 - 7.45

Specification Test Results

Frequently Bought Together