À propos de cet article
BioClean RABS and isolator sleeves are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties.
- Ultra-clean surface ensures product protection
- 100% inspected and air leak tested (prior to being guillotined)
- Sterilised via gamma irradiation
- 90 mm cuff length
Designed for use in product contact areas, BioClean RABS/Isolator sleeves are fully validated for sterility with an SAL (Sterility Assurance Level) of 10⁻⁶ and have a cuff ring diameter of 90 mm to fit most available cuff ring systems.
Suitable for use in ISO Class 4 and EU GMP Grade A environments. Three years from the date of manufacture. Store in a dry, cool place away from sunlight and florescent light.
Food contact approved.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Catalog No:
- NITRGSL36NITPP26
- NITRGSL15NITPP26
- NITRGSL20NITPP26
- NITRGSL33NITPP26
- Anti-static:
- +
- +
- +
- +
- Cleanroom class:
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- Garment application:
- Cleanroom, Medical, Laboratory, Clinical, Electronics/semi-conductor
- Cleanroom, Medical, Laboratory, Clinical, Electronics/semi-conductor
- Cleanroom, Medical, Laboratory, Clinical, Electronics/semi-conductor
- Cleanroom, Medical, Laboratory, Clinical, Electronics/semi-conductor