Order Entry
Belgium
ContactUsLinkComponent
 
Test paper, qualitative, wator
Catalog # MANA90610
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test paper, qualitative, wator
Catalog # MANA90610
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Wetting of a hydrophilic paper
  • Pk:
    50
  • Description:
    Wator, for water distribution in butter
  • Tests per kit:
    50 sheets 78×40 mm

Specifications

About this item

The test paper allows the quick and easy determination of the water distribution in butter. It is a special presentation of Watesmo for the dairy industry.

  • On contact with water the paper develops dark spots

Size and number of these spots are correlated to the distribution of water in butter. This is evaluated with a five point evaluation chart described in DIN 10 311. For water detection for other applications we recommend Watesmo.

Colour change: Light blue to dark blue