1-Butanesulphonic acid sodium salt ≥99.0% (dried basis), HiPerSolv CHROMANORM® for HPLC
85790.150
:
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
- Assay (calculated on dried substance):≥ 99,0 %
- Appearance :White powder
- IR Spectrum :Passes test
- Appearance of solution (10 %):Clear and colourless
- Water :≤ 1,0 %
- Absorbance (210 nm) (0.5 mol/l) :≤ 0,1
- Absorbance (220 nm) (0.5 mol/l) :≤ 0,06
- Absorbance (230 nm) (0.5 mol/l) :≤ 0,04
- Absorbance (260 nm) (0.5 mol/l) :≤ 0,02
:
:160.17 g/mol
:Ambient
:2386-54-1
:C₄H₉NaO₃S
:MFCD00007540