Order Entry
Belgium
ContactUsLinkComponent
ASTM® D6584 1,2,4-Butanetriol solution, 1000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
ASTM® D6584 1,2,4-Butanetriol solution, 1000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
Catalog # HONCR1384-5ML
CAS Number:  
ASTM® D6584 1,2,4-Butanetriol solution, 1000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
Catalog # HONCR1384-5ML
CAS Number:  

Specifications

  • Concentration:
    1000 μg/ml in pyridine
  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Description:
    ASTM® D6584 1,2,4-Butanetriol solution
  • Storage temperature:
    2...8 °C
  • Description:
    ASTM® D6584 1,2,4-Butanetriol solution
  • Flash point:
    20 °C
  • Boiling point:
    115 °C
  • Freezing point:
    –42 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results