Pesticide reference standard, Amicarbazone, neat, Honeywell Fluka™
Catalog # HONCR1335-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:129909-90-6
- Description:Amicarbazone
- Environmentally Preferable:
- Formula:C₁₀H₁₉N₅O₂
- Storage temperature:Ambient
- Description:Amicarbazone
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results