Order Entry
Belgium
ContactUsLinkComponent
 
Pesticide reference standard, Flucarbazone sodium, neat, Honeywell Fluka™
  HONCR1310-100MG
 :  
Pesticide reference standard, Flucarbazone sodium, neat, Honeywell Fluka™
  HONCR1310-100MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • CAS No.:
    181274-17-9
  • Description:
    Flucarbazone sodium
  • Environmentally Preferable:
  • Formula:
    C₁₂H₁₀F₃N₄NaO₆S
  • Storage temperature:
    Ambient
  • Description:
    Flucarbazone sodium
  • Freezing point:
    200 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results