Order Entry
Belgium
ContactUsLinkComponent
 
Pesticide reference standard, Ipfencarbazone, neat, Honeywell Fluka™
Catalog # HONCR1308-25MG
CAS Number:  
Pesticide reference standard, Ipfencarbazone, neat, Honeywell Fluka™
Catalog # HONCR1308-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mg
  • CAS No.:
    212201-70-2
  • Description:
    Ipfencarbazone
  • Environmentally Preferable:
  • Formula:
    C₁₈H₁₄Cl₂F₂N₄O₂
  • Storage temperature:
    Ambient
  • Description:
    Ipfencarbazone
  • Freezing point:
    129...134 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results