Pesticide reference standard, Sulfentrazone, neat, Honeywell Fluka™
Catalog # HONCR1066-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:122836-35-5
- Description:Sulfentrazone
- Environmentally Preferable:
- Formula:C₁₁H₁₀Cl₂F₂N₄O₃S
- Storage temperature:Ambient
- Description:Sulfentrazone
- Freezing point:122 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results