ContactUsLinkComponent
Quantitative filter papers, hardened ashless grades, grade 391
Quantitative filter papers, hardened ashless grades, grade 391
Leverancier:  Ahlstrom-Munksjö
Certificaten
undefined
Quantitative filter papers, hardened ashless grades, grade 391
Leverancier:  Ahlstrom-Munksjö
Specificaties
  • Catalog No:
  • BINZ3.104.300
  • BINZ3.104.012
  • BINZ3.104.006
  • BINZ3.104.042-5
  • BINZ3.104.042
  • BINZ3.104.500
  • BINZ3.104.400
  • Grade:
  • 391
  • 391
  • 391
  • 391
  • 391
  • 391
  • 391
  • Particle retention:
  • 2 - 3 μm
  • 2 - 3 μm
  • 2 - 3 μm
  • 2 - 3 μm
  • 2 - 3 μm
  • 2 - 3 μm
  • 2 - 3 μm
  • Filtration speed:
  • 180
  • 180
  • 180
  • 180
  • 180
  • 180
  • 180
  • Basis weight:
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
Specificaties
Frequently Bought Together