ContactUsLinkComponent
Wipes for delicate tasks, VERACLEAN® I-Tack
Wipes for delicate tasks, VERACLEAN® I-Tack
Catalog # PGIN74236
Supplier:  PGI NONWOVEN
undefined
Wipes for delicate tasks, VERACLEAN® I-Tack
Catalog # PGIN74236
Supplier:  PGI NONWOVEN
Specifications
  • Packed:
    25 sheets/pack, 16 packs/case
  • Colour:
    Green
  • Description:
    Wipes, VERACLEAN® I-Tack, ¹/₄ fold
  • L×W:
    610×370 mm
  • Pk:
    400
Specifications
About this item
These wipes are specially designed to swiftly and cleanly remove contaminants prior to painting. This I-Tack range contains a water-based tackifier and is compatible with all paint types.
  • The cloths are cost efficient because they can be used up to eight sides
  • No unpleasant smells or sticky residue on either your hands or the car body