Order Entry
Belgium
ContactUsLinkComponent
 
Lactophenol blue solution stain (fungal), Sigma-Aldrich®
Catalog # 1.13741.0100
Supplier:  Merck
CAS Number:  
undefined
Lactophenol blue solution stain (fungal), Sigma-Aldrich®
Catalog # 1.13741.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Pk Info:
    Preparation Capacity 100 slides

Specifications

About this item

 :
Density:1.16 g/cm3 (20 °C)
UN:2821
ADR:6.1,
 :M-Clarity™ quality level = MQ300
 :GHS05
 :GHS08
 :GHS07
Storage Temperature:Ambient
 :1
MDL Number:MFCD00146574