Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Copper
VISOCOLOR® Colorimetric Test, Copper
   Sustainable Options Available
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Copper
Supplier:  MACHEREY-NAGEL
   Sustainable Options Available
Certificates
VISOCOLOR® Colorimetric Test, Copper
4 Options Available Below
Product Details & Documents
Reagents for the determination of copper in water samples.
  • Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: No
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • MANA931037
  • MANA920050
  • MANA931237
  • MANA920150
  • Platform:
  • Colorimetric test kits
  • Colorimetric test kits
  • Colorimetric test kits
  • Colorimetric test kits
  • Storage temperature:
  • 15...25 °C
  • 15...25 °C
  • 15...25 °C
  • 15...25 °C
  • Shelf life (months):
  • 24
  • 24
  • 24
  • 24
  • Weight:
  • 221,87 g (Including packaging)
  • 520 g (Including packaging)
  • 131,8 g (Including packaging)
  • 276,02 g (Including packaging)
Specifications
Product Family Options
Test kits
Product Information
  • Contents
    Range
    Chemical method
    For unit
    Dimensions
    Tests per kit
    Pk
    Availability
    Price
  • MANA931037
    VISOCOLOR® ECO Copper, test no. 5-37; Measuring range on PF-12Plus. Range on other photometers can be different.
    VISOCOLOR® ECO Copper, test no. 5-37; Measuring range on PF-12Plus. Range on other photometers can be different.
    Reagents for 100 tests, color chart and accessories in a box
    0 • 0,1 • 0,2 • 0,3 • 0,5 • 0,7 • 1,0 • 1,5 mg/L (Cu²⁺); 0,1 - 5,0 mg/L (Cu²⁺)
    Cuprizon
    PF-12Plus;PF-3 Fish;Advance;Visual
    168×112×40 mm (Including packaging)
    100
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 65,60 €
    65,60 €
    1 * 1 KIT
       65,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA931037
    Platform
    Colorimetric test kits
    Storage temperature
    15...25 °C
    Shelf life (months)
    24
    Weight
    221,87 g (Including packaging)
  • MANA920050
    VISOCOLOR® HE Copper
    VISOCOLOR® HE Copper
    Reagents for 150 tests, color chart and accessories in a box
    0,0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0.25 • 0,30 • 0,40 • 0,50 mg/L (Cu²⁺)
    Cuprizon
    Visual
    245×145×47 mm (Including packaging)
    150
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 124,00 €
    124,00 €
    1 * 1 KIT
       124,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA920050
    Platform
    Colorimetric test kits
    Storage temperature
    15...25 °C
    Shelf life (months)
    24
    Weight
    520 g (Including packaging)
Refill packs
Product Information
  • Contents
    Range
    Chemical method
    For unit
    Dimensions
    Tests per kit
    Pk
    Availability
    Price
  • MANA931237
    VISOCOLOR® ECO Copper, refill pack, test no. 5-37; Measuring range on PF-12Plus. Range on other photometers can be different.
    VISOCOLOR® ECO Copper, refill pack, test no. 5-37; Measuring range on PF-12Plus. Range on other photometers can be different.
    Reagents for 100 tests in a box
    0 • 0,1 • 0,2 • 0,3 • 0,5 • 0,7 • 1,0 • 1.5 mg/L (Cu²⁺); 0,1 - 5,0 mg/L (Cu²⁺)
    Cuprizon
    PF-12Plus;PF-3 Fish;Advance;Visual
    170×115×70 mm (Including packaging)
    100
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 36,70 €
    36,70 €
    1 * 1 KIT
       36,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA931237
    Platform
    Colorimetric test kits
    Storage temperature
    15...25 °C
    Shelf life (months)
    24
    Weight
    131,8 g (Including packaging)
  • MANA920150
    VISOCOLOR® HE Copper, refill pack
    VISOCOLOR® HE Copper, refill pack
    Reagents for 150 tests in a box
    0,0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 • 0,50 mg/L (Cu²⁺)
    Cuprizon
    Visual
    233×129×45 mm (Including packaging)
    150
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 76,80 €
    76,80 €
    1 * 1 KIT
       76,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA920150
    Platform
    Colorimetric test kits
    Storage temperature
    15...25 °C
    Shelf life (months)
    24
    Weight
    276,02 g (Including packaging)
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.