Orders LinkContactUsLinkComponent
Face shield, Turboshield™
Face shield, Turboshield™
Supplier:   HONEYWELL SAFETY
Face shield, Turboshield™
Supplier:  HONEYWELL SAFETY
Certificates
Face shield, Turboshield™
3 Options Available Below
Product Details & Documents
The Turboshield™ is a great value faceshield that offers easy visor exchange, is more comfortable to wear and offers better protection.
  • Simple push‐button release system allows for easy visor exchange in seconds
  • Breathable, removable, washable headband
  • Toric lens provides excellent optics, increased field of view and added chin coverage without bulky attachments
  • Ergonomic, easy‐to‐use adjustment knobs
Applications include grinding, welding, chemical, construction, food services, manufacturing, medical, oil and gas, pharmaceutical, steel and metals.
Certifications: AS/NZS 1337.1:2010
Product Family Options
Product Information
  • Type
    Material
    L×W
    Availability
    Price
  • Chemical and impact resistant
    PC
    240x 230 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,40 €
    22,40 €
    1 * 1 ST
       22,40 €
  • Chemical and impact resistant
    PC
    240x 230 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,20 €
    51,20 €
    1 * 1 ST
       51,20 €
  • Chemical and impact resistant
    240x 230 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,30 €
    51,30 €
    1 * 1 ST
       51,30 €