Order Entry
Belgium
ContactUsLinkComponent
 
Digital multispeed vortex mixer, MSV-3500
Catalog # GRANMSV-3500
Supplier:  GRANT INSTRUMENTS
undefined
Digital multispeed vortex mixer, MSV-3500
Catalog # GRANMSV-3500
Supplier:  GRANT INSTRUMENTS

Specifications

  • Max. capacity:
    0,2...50 ml
  • Orbit:
    4 mm
  • Shaking motion:
    Orbital
  • Speed range:
    300...3500 min⁻¹
  • Description:
    Digital Mutispeed vortex mixer, MSV-3500
  • Model:
    MSV-3500
  • Power supply:
    120 - 230 V
  • Weight:
    2,6 kg
  • W×D×H:
    1800×1700×1450 mm
  • Pk:
    1

Specifications

About this item

The Grant bio MSV-3500 is a versatile multi speed vortex mixer is designed for soft or intensive mixing of reagents in different size and type plastic tubes. It is designed for operation in life-science laboratories working in the fields of biochemistry, cell and molecular biology.

  • Four types of interchangeable platforms
  • Microprocessor control, LCD display
  • Acceleration time to max speed: 15 seconds

Provides high maximum speed of platform rotation efficiently mixing microvolumes (less than 5 ml) of samples.

Widely used in intensive stirring of bacterial and yeast cells, extraction of metabolites and enzymes from cells and cell cultures, and various DNA operations such as deproteinisation of DNA/protein complexes and purification of low molecular weight DNA/RNA fragments in PCR diagnostics.

Delivery: Platforms are sold separately or available as part of a kit (GRANMSV-3500 4P).