Order Entry
Belgium
ContactUsLinkComponent
Calbiochem® Mouse Albumin
Calbiochem® Mouse Albumin
Catalog # 126674-25
CAS Number:  
Calbiochem® Mouse Albumin
Catalog # 126674-25
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    25 mg
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Form:
    Lyophilised
  • Protein synonyms:
    MSA
  • Protein/peptide name:
    Albumin
  • Purity:
    >98%

Specifications

About this item

Plasma protein used as a stabilising agent and a carrier protein.