Orders LinkContactUsLinkComponent
Urine containers, SpecTainer™ II
Urine containers, SpecTainer™ II
Supplier:   Simport Scientific
Urine containers, SpecTainer™ II
Supplier:  Simport Scientific
Certificates
Urine containers, SpecTainer™ II
Urine containers, SpecTainer™ II
2 Options Available Below
Product Details & Documents
PP urine container with leak resistant PE screw cap, ideal for collecting, storing and transporting urine samples.
  • Graduated, except 60 ml container
  • Ridges around base offer a better grip during opening and closing
  • Stackable, shatter resistant
  • Designed with a straight side format
  • Moulded ridges around lid make it easy to open and close
Warning label has space for patient identification. For colour coding purposes, place a Capinsert™ on top of closure (see Series T345).
Product Family Options
With yellow cap
Product Information
  • Capacity
    Version
    Availability
    Price
  • 60 ml
    Non sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 438,00 €
    438,00 €
    1 * 500 ST
       438,00 €
With cyan cap
Product Information
  • Capacity
    Version
    Availability
    Price
  • 90 ml
    Sterile
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 516,00 €
    516,00 €
    1 * 400 ST
       516,00 €