Order Entry
Belgium
ContactUsLinkComponent
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) 10 mg/mL in water
Irritant
Health
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) 10 mg/mL in water
Catalog # 40046.
Supplier:  Biotium
CAS Number:  
undefined
Irritant
Health
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) 10 mg/mL in water
Catalog # 40046.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :561.93 g/mol
EINECS:245-691-1
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:23491-52-3
 :MFCD00012679
Formula:C₂₇H₂₈N₆O·3HCl