Order Entry
Belgium
ContactUsLinkComponent
Propidium iodide 50 ug/mL in PBS
Propidium iodide 50 ug/mL in PBS
Catalog # 40048.
Supplier:  Biotium
CAS Number:  
Propidium iodide 50 ug/mL in PBS
Catalog # 40048.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :668.4 g/mol
Storage Temperature:Refrigerator
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921