Order Entry
Belgium
ContactUsLinkComponent
Propidium iodide 1 mg/ml in water
Propidium iodide 1 mg/ml in water
Catalog # 40017.
Supplier:  Biotium
CAS Number:  
undefined
Propidium iodide 1 mg/ml in water
Catalog # 40017.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :668.4 g/mol
Storage Temperature:Refrigerator
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921