Orders LinkContactUsLinkComponent
Wipes, Amplitude™ Sigma™ Plus
Wipes, Amplitude™ Sigma™ Plus
Supplier:   CONTEC
Wipes, Amplitude™ Sigma™ Plus
Supplier:  CONTEC
Certificates
Wipes, Amplitude™ Sigma™ Plus
Wipes, Amplitude™ Sigma™ Plus
2 Options Available Below
Product Details & Documents
The Amplitude™ Sigma™ Plus wipes are strong and durable, compatible with most solvents, and have outstanding sorbency.
  • Hydroentangled cellulose/poylester provides excellent strength and durability
  • Low levels of particles and extractables
  • Variety of blended fibers enable excellent absorbing capacity
Amplitude™ Sigma™ Plus Wipes are constructed using a nonwoven fabric that is a hydroentangled blend of polyester and cellulose fibers. Wet or dry, the wipe maintains a high level of strength and durability.
Specially treated to reduce particles and fibers, it is recommended when low particle generation combined with sorbency is of high priority.
Product Family Options
Product Information
  • Colour
    L×W
    Availability
    Price
  • CONTC1E-99
    Amplitude™ Sigma™ Plus wipes, flat stacked
    Amplitude™ Sigma™ Plus wipes, flat stacked
    White
    229×229 mm
    Log in for availability
    Log in for availability
    1 * 3.600 ST
    1 * 3.600 ST - 856,00 €
    856,00 €
    1 * 3.600 ST
       856,00 €
  • CONTC1E-1212
    Amplitude™ Sigma™ Plus wipes, flat stacked
    Amplitude™ Sigma™ Plus wipes, flat stacked
    White
    305×305 mm
    Log in for availability
    Log in for availability
    1 * 2.700 ST
    1 * 2.700 ST - 1 320,00 €
    1 320,00 €
    1 * 2.700 ST
       1 320,00 €