Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 1.0 N in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Corrosive
Sodium hydroxide concentrate for dilution to 1.0 N in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # 4693-09
CAS Number:  
Corrosive
Sodium hydroxide concentrate for dilution to 1.0 N in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # 4693-09
CAS Number:  

Specifications

  • Pack type:
    Cubetainer®
  • Pk:
    20 L
  • Seq:
    5302

Specifications

Specification Test Results

  • Normality:
    0.995 - 1.005

Specification Test Results

About this item

 :
 :40 g/mol
Density:1.04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548