Orders LinkContactUsLinkComponent
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
Supplier:   MOTIC
FLED fluorescence intermediate attachments
Supplier:  MOTIC
Certificates
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
3 Options Available Below
Product Details & Documents
Compatible with Panthera U, KU and CC microscopes.
Product Family Options
Product Information
  • Availability
    Price
  • MOTI1101011400181
    FLED Fluorescence intermediate attachment (Auramine O)
    - Fluorescence intermediate part
    - LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
    - Light blocking slider
    Note: Recommended to use with Plan objectives UC PL NCG
    FLED Fluorescence intermediate attachment (Auramine O)
    - Fluorescence intermediate part
    - LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
    - Light blocking slider
    Note: Recommended to use with Plan objectives UC PL NCG
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 580,00 €
    2 580,00 €
    1 * 1 ST
       2 580,00 €
  • MOTI1101011400191
    FLED Fluorescence intermediate attachment (FITC)
    - Fluorescence intermediate part
    - LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
    - Light blocking slider
    FLED Fluorescence intermediate attachment (FITC)
    - Fluorescence intermediate part
    - LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
    - Light blocking slider
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 250,00 €
    2 250,00 €
    1 * 1 ST
       2 250,00 €
  • MOTI1101011400201
    FLED Fluorescence intermediate attachment (TRITC)
    - Fluorescence intermediate part
    - LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
    - Light blocking slider
    FLED Fluorescence intermediate attachment (TRITC)
    - Fluorescence intermediate part
    - LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
    - Light blocking slider
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 250,00 €
    2 250,00 €
    1 * 1 ST
       2 250,00 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.