Order Entry
Belgium
ContactUsLinkComponent
Sodium sulphate, anhydrous ≥99.0%, granular ACS, Fluka™
Sodium sulphate, anhydrous ≥99.0%, granular ACS, Fluka™
Certificates
undefined
Sodium sulphate, anhydrous ≥99.0%, granular ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Loss on ignition (800°C):
    Max. 0.5 %
  • Water insoluble matter:
    Max. 0.01 %
  • pH (5 %, 25 °C):
    5.2 - 9.2
  • Calcium (Ca):
    Max. 0.01 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 10 ppm
  • Phosphate (PO₄):
    Max. 0.001 %
  • Total N:
    Max. 5 ppm
  • Grain Size 10 - 60 Mesh:
    Min. 85 %

Specification Test Results

Related Information

Frequently Bought Together