Sodium molybdate(VI) dihydrate ≥99% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate
CAS Number:10102-40-6
:
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170
Density:2.700 g/cm³ (20 °C)
Molecular Weight:241.95 g/mol
Melting Pt:100 °C (–H₂O of crystall.)
Merck Index:13,08718
Storage Temperature:Ambient
EINECS:231-551-7
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.5 - 103.0 %
- pH (5 %, 25 °C):7.0 - 10.5
- Water insoluble matter:Max. 0.005 %
- Iron (Fe):Max. 0.001 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 0.005 %
- Ammonium (NH₄):Max. 0.001 %
- Phosphate (PO₄):Max. 5 ppm
- Sulfate (SO₄):Max. 0.015 %
Specification Test Results
Frequently Bought Together