Order Entry
Belgium
ContactUsLinkComponent
 
Sodium chloride 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP (max. 0.00002% Al), Fluka™
 
Sodium chloride 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP (max. 0.00002% Al), Fluka™
 

 

  • Assay (calculated to the dried substance):
    99.0 - 100.5 %
  • Loss on drying (105°C, 2 h):
    Max. 0.5 %
  • Acidity or alkalinity:
    complying
  • Aluminium (Al):
    Max. 0.2 ppm
  • Arsenic (As):
    Max. 1 ppm
  • Barium (Ba):
    complying
  • Iron (Fe):
    Max. 2 ppm
  • Potassium (K):
    Max. 500 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Bromide (Br):
    Max. 0.005 %
  • Iodide (I) test:
    complying
  • Ferrocyanides:
    complying
  • Nitrite (NO₂):
    complying
  • Phosphate (PO₄):
    Max. 25 ppm
  • Sulfate (SO₄):
    Max. 200 ppm
  • Appearance of the solution:
    complying
  • Mg, earth alkali metals (as Ca):
    Max. 100 ppm
  • Identity:
    complying
  • Residual solvents:
    complying

 

 

Frequently Bought Together