Order Entry
Belgium
ContactUsLinkComponent
Potassium iodide concentrate for dilution to 1L, 0.1 M KI volumetric solution, Fluka™
Health
Potassium iodide concentrate for dilution to 1L, 0.1 M KI volumetric solution, Fluka™
Catalog # 38110-1EA
CAS Number:  
Health
Potassium iodide concentrate for dilution to 1L, 0.1 M KI volumetric solution, Fluka™
Catalog # 38110-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :166 g/mol
 :GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405