Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate 1/6 M K2Cr2O7 (1N) volumetric solution, Fluka™
Health
Corrosive
Irritant
Potassium dichromate 1/6 M K2Cr2O7 (1N) volumetric solution, Fluka™
Catalog # 34271-1L
CAS Number:  
Health
Corrosive
Irritant
Potassium dichromate 1/6 M K2Cr2O7 (1N) volumetric solution, Fluka™
Catalog # 34271-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :294.18 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.030 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3287
ADR:6.1,III
 :GHS08
 :GHS05
 :GHS07
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367