Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Irritant
Health
N-Methyl-2-pyrrolidone (NMP) ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Catalog # 69337-6X1L
CAS Number:  
undefined
Irritant
Health
N-Methyl-2-pyrrolidone (NMP) ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Catalog # 69337-6X1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    6 L
  • Seq:
    5198

Specifications

Specification Test Results

  • Refractive Index:
    n20/D 1.470
  • Assay (GC):
    Min. 99.9 %
  • Refractive index (n 20/D):
    1.469 - 1.471
  • Water (Karl Fischer):
    Max. 0.1 %
  • Absorbance at 285 nm:
    Max. 0.522
  • Absorbance at 300 nm:
    Max. 0.259
  • Absorbance at 320 nm:
    Max. 0.107
  • Absorbance at 350 nm:
    Max. 0.013
  • Suitability for the GC Headspace:
    complying
  • Explosion Limit:
    9.5 %
  • Autoignition Temperature:
    518 °F

Specification Test Results

About this item

 :
 :99.13 g/mol
Melting Pt:-24 °C
Density:1.0259 g/cm3 (25 °C)
Flash Pt:88 °C
EINECS:212-828-1
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193
Melting Pt:–24 °C