
Lithium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™
:7447-41-8
:
:LiCl
:MFCD00011078
:42.39 g/mol
:1.360 °C (1.013 hPa)
:605 °C
:2.07 g/cm³ (20 °C)
:Ambient
:231-212-3
:GHS07
- Assay:Min. 99.0 %
- Water insoluble matter:Max. 0.01 %
- Loss on drying (105°C, 2 h):Max. 1.0 %
- Titrable Base:Max. 0.008 meq/g
- Barium (Ba):Max. 0.003 %
- Calcium (Ca):Max. 0.01 %
- Iron (Fe):Max. 0.001 %
- Potassium (K):Max. 0.01 %
- Sodium (Na):Max. 0.20 %
- Heavy metals (ICP) ACS:Max. 0.002 %
- Nitrate (NO₃):Max. 0.001 %
- Sulfate (SO₄):Max. 0.01 %
Frequently Bought Together