Order Entry
Belgium
ContactUsLinkComponent
Cesium chloride, anhydrous ≥99.9%, Reagent Grade, free-flowing, Fluka™
Health
Cesium chloride, anhydrous ≥99.9%, Reagent Grade, free-flowing, Fluka™
Certificates
Health
Cesium chloride, anhydrous ≥99.9%, Reagent Grade, free-flowing, Fluka™

Specification Test Results

  • Assay:
    99 - 101 %
  • Metal trace analysis (ICP):
    Max. 1500 ppm

Specification Test Results

Related Information

Frequently Bought Together