Order Entry
Belgium
ContactUsLinkComponent
2-Propanol ≥99.9% (by GC), TraceSELECT™ for metal speciation analysis, Riedel-de Haën™
Irritant
Flammable
2-Propanol ≥99.9% (by GC), TraceSELECT™ for metal speciation analysis, Riedel-de Haën™
Catalog # 04516-1L
CAS Number:  
undefined
Irritant
Flammable
2-Propanol ≥99.9% (by GC), TraceSELECT™ for metal speciation analysis, Riedel-de Haën™
Catalog # 04516-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Silver (Ag):
    Max. 0.5 ppb
  • Aluminium (Al):
    Max. 5 ppb
  • Arsenic (As):
    Max. 1 ppb
  • Gold (Au):
    Max. 0.5 ppb
  • Barium (Ba):
    Max. 0.5 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • Bismuth (Bi):
    Max. 0.5 ppb
  • Calcium (Ca):
    Max. 30 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 0.5 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 1 ppb
  • Iron (Fe):
    Max. 5 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Mercury (Hg):
    Max. 1 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Iridium (Ir):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 10 ppb
  • Manganese (Mn):
    Max. 5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 250 ppb
  • Nickel (Ni):
    Max. 5 ppb
  • Lead (Pb):
    Max. 0.5 ppb
  • Palladium (Pd):
    Max. 1 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 0.5 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Tin (Sn):
    Max. 0.5 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 5 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Uran (U):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 0.5 ppb
  • Zinc (Zn):
    Max. 5 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Bromide (Br):
    Max. 10 ppb
  • Chloride (Cl):
    Max. 25 ppb
  • Fluoride (F):
    Max. 10 ppb
  • Iodide (I):
    Max. 10 ppb
  • Nitrite (NO₂):
    Max. 10 ppb
  • Nitrate (NO₃):
    Max. 25 ppb
  • Phosphate (PO₄):
    Max. 50 ppb
  • Sulfate (SO₄):
    Max. 50 ppb
  • Peroxides (as H₂O₂):
    Max. 5 ppm
  • Propionaldehyde (GC):
    Max. 20 ppm
  • Ketones (as CH₃COCH₃):
    Max. 20 ppm
  • Transmittance from 260 nm:
    Min. 98 %
  • Transmittance at 230 nm:
    Min. 75 %
  • Transmittance at 220 nm:
    Min. 50 %
  • Autoignition Temperature:
    750 °F

Specification Test Results

About this item

 :
 :60.1 g/mol
Boiling Pt:77…83 °C (1.013 hPa)
Melting Pt:–90 °C
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :GHS07
 :GHS02
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674