Order Entry
Belgium
ContactUsLinkComponent
2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Flammable
Health
Irritant
2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates
undefined
Flammable
Health
Irritant
2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates

Specification Test Results

  • Assay (GC):
    Min. 99.0 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Absorbance at 270 nm:
    Max. 0.01
  • Absorbance at 235 nm:
    Max. 0.10
  • Absorbance at 210 nm:
    Max. 0.70

Specification Test Results

Related Information

Frequently Bought Together