Order Entry
Belgium
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Chloride

QUANTOFIX® Test Strips, Chloride

QUANTOFIX® carbonate hardness, 0 • 4 • 8 • 13 • 19 • 25 - °e, Reaction of an acid in the presence of a mix indicator, 43×123×43 mm (packaging), 100 Tests, 100 Tests
Catalog # MANA91323
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Chloride
Catalog # MANA91323
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Chloride
QUANTOFIX® Test Strips, Chloride

QUANTOFIX® carbonate hardness, 0 • 4 • 8 • 13 • 19 • 25 - °e, Reaction of an acid in the presence of a mix indicator, 43×123×43 mm (packaging), 100 Tests, 100 Tests

CAS Number:  
Price
34,70 €
Log in for availability
1 * 1 EA
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips are for the rapid and reliable determination of chloride in solutions. The easy dip-and-read-procedure provides a reliable result within 1 minute.

Specifications

Specifications

  • Chemical method:
    Reaction of an acid in the presence of a mix indicator
  • Pk:
    100 Tests
  • Description:
    QUANTOFIX® carbonate hardness
  • Dimensions:
    43×123×43 mm (packaging)
  • Tests per kit:
    100 Tests
  • Range:
    0 • 4 • 8 • 13 • 19 • 25 - °e
  • Cat. No.:
    MANA91323
  • Parameters:
    All reagents included
  • Contents:
    100 test strips in a tube, instruction leaflet
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    No
  • Shelf life (months):
    30 (from production)
  • Method:
    Carbonate hardness - Mixed indicator (bright green → blue)
  • Weight:
    37 g

Specifications