Order Entry
Belgium
ContactUsLinkComponent
Test strips, ozone
Test strips, ozone
Catalog # MANA90736
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, ozone
Catalog # MANA90736
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Ozone test strips
  • For unit:
    26128534
  • Tests per kit:
    12
  • Range:
    90 - 210 µg/m3 O₃

Specifications

About this item

These Ozone test strips are designed for quick, convenient and reliable on-site Ozone testing. The easy procedure yields accurate testing results within ten minutes.

  • Results within minutes: Just expose to air, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

Ozone is a colourless toxic gas. It is irritating to eyes and mucous membranes and can cause respiratory complaints. With Ozone concentrations increasing world-wide, especially in the summer time, Ozone testing becomes more and more important.

An Ozone content of 180 μg/m³ in the air is considered as critical value above which sensitive persons should avoid strong physical exertion. The Ozone values published in the media only represent the Ozone concentrations in the vicinity of the measuring stations. However, actual Ozone values in other locations can deviate considerably from these published values due to meteorological and chemical influences. The reasons for such deviations are for wind, solar radiation, exhaust gases from vehicles and so on. For this reason an on-site measurement of Ozone concentrations is recommended.

Colour reaction: White to brown