


α-Naphthyl Acetate (Non-Specific Esterase) Kit Kit formulated with all liquid reagents., Sigma-Aldrich®
Catalog # 91A-1KT
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
About this item
:
ADR:9,II
:M-Clarity™ quality level = MQ500 (more info.)
Flash Pt:36 °C
UN:3316
:GHS02
:GHS06
:GHS08
Storage Temperature:Refrigerator
:1
MDL Number:MFCD08705438