Sample storage tubes, blank and alphanumeric
About this item
Abgene blank and alphanumeric storage tubes are manufactured from 100% virgin, medical-grade polypropylene. Storage tubes are leak tested to ensure the integrity and security of samples and with a V-bottom design, our alphanumeric storage tubes facilitate sample recovery.
- Highest quality
- Enhanced rack design
Unlike traditional storage tubes or blocks, our blank and alphanumeric storage tubes are available in our specially-designed, stackable, 96-format, microplate-footprint racks to save precious space in your freezer, refrigerator, compound store or on the bench top. In addition, the rack design includes grid referenced lids for simple identification of tube location.
4 Options Available Below
Product Details & Documents
Abgene blank and alphanumeric storage tubes are manufactured from 100% virgin, medical-grade polypropylene. Storage tubes are leak tested to ensure the integrity and security of samples and with a V-bottom design, our alphanumeric storage tubes facilitate sample recovery.
- Highest quality
- Enhanced rack design
Unlike traditional storage tubes or blocks, our blank and alphanumeric storage tubes are available in our specially-designed, stackable, 96-format, microplate-footprint racks to save precious space in your freezer, refrigerator, compound store or on the bench top. In addition, the rack design includes grid referenced lids for simple identification of tube location.
Specifications for Product Family
Specifications
- Catalog No:
- 732-0495
- 732-4827
- 732-4845
- 732-4919
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
- CapacityPackedTypeAvailabilityPrice
- 732-0495Microtube cluster rack with lidMicrotube cluster rack with lid0,65 ml10 racks of 96 tubes/caseBlank storage tubeLog in for availability1 * 960 ST560,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.732-0495Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 732-4827Microtubes in a cluster rack with lid, each rack containing 96 microtubesMicrotubes in a cluster rack with lid, each rack containing 96 microtubes1,2 ml10 racks of 96 tubes/caseBlank storage tubeLog in for availability1 * 960 ST577,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.732-4827Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 732-4845Microtubes tubes (loose)Microtubes tubes (loose)1,2 mlBulk, 1000 tubes/caseBlank storage tubeLog in for availability1 * 1.000 ST145,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.732-4845Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 732-4919Microtubes in grid-referenced microtube cluster rackMicrotubes in grid-referenced microtube cluster rack1,2 ml10 racks of 96 tubes/caseAlphanumeric tubeLog in for availability1 * 960 ST673,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.732-4919Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact