ContactUsLinkComponent
MS spent media analysis columns, AdvanceBio
MS spent media analysis columns, AdvanceBio
Supplier:  Agilent
Certificates
undefined
MS spent media analysis columns, AdvanceBio
Supplier:  Agilent
Specifications
  • Catalog No:
  • HEWL675775-901
  • HEWL673775-901
  • HEWL679775-901
  • Max. pressure:
  • 600 bar
  • 600 bar
  • 600 bar
  • pH range:
  • 3,0 – 11,0
  • 3,0 – 11,0
  • 3,0 – 11,0
  • Temperature limit:
  • 80 °C
  • 80 °C
  • 80 °C