Handheld digital microscopes, Dino-Lite Wireless
Leverancier: Dino-Lite / IDCP B.V.
Certificaten
Over dit artikel
These microscopes from the Dino-Lite Universal series work wired via USB or wireless with the integrated WF-20 Wireless adapter.
- Wireless and wired in one device
- Work with different operating systems: iOS, Android or Windows
- USB 2.0, enhanced 1,3 MP EDGE sensor
- Integrated polariser greatly reduces glare on reflective surfaces
- Extensive measurement functionality
- Flexible LED control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Ideal for inspections in hard-to-reach places in factories, in the field, in workshops or in greenhouses.
Gerelateerde informatie
Frequently Bought Together