Overhead stirrer tools
About this item
These high-speed overhead stirrer tools are designed for mixing applications in the laboratory specialised for low- to medium-viscosity materials.
- Jog-Shuttle controller with touch-button
- Maximal viscosity 10000/50000 mPas
- Chrome-plated stainless-steel chuck to prevent corrosion
- Impeller height adjustable
- Six light sources and LCD display
These units provide steady and constant stirring speed and designed with powerful DC motor for various stirring applications such as direct-driven or worm-geared.
The direct driven motor ensures high-speed at all recommended viscosities. Features with anti-trembling system that is the impeller is fixed at two points within the body and chuck.
These units are features with timer (99 h 59 min) with timer-end signal, locking mode, error status display, overload and overheat protection and operation LEDs.
: Available as set with impeller, plate stand and clamp, many different impellers and plate stands available.
3 Options Available Below
Product Details & Documents
These high-speed overhead stirrer tools are designed for mixing applications in the laboratory specialised for low- to medium-viscosity materials.
- Jog-Shuttle controller with touch-button
- Maximal viscosity 10000/50000 mPas
- Chrome-plated stainless-steel chuck to prevent corrosion
- Impeller height adjustable
- Six light sources and LCD display
These units provide steady and constant stirring speed and designed with powerful DC motor for various stirring applications such as direct-driven or worm-geared.
The direct driven motor ensures high-speed at all recommended viscosities. Features with anti-trembling system that is the impeller is fixed at two points within the body and chuck.
These units are features with timer (99 h 59 min) with timer-end signal, locking mode, error status display, overload and overheat protection and operation LEDs.
: Available as set with impeller, plate stand and clamp, many different impellers and plate stands available.
Product Family Options
Product Information
- Shaft ØHeightWidthLengthMax. revolutionsAvailabilityPrice
- WITGDH.WOS01016Overhead stirrer, HS-30 D set, 10 L, 230 VOverhead stirrer, HS-30 D set, 10 L, 230 V8 mm180 mm90 mm145 mm200 - 3000 min-1Log in for availability1 * 1 ST737,00 €
- WITGDH.WOS01025Overhead stirrer, HS-100 D unit, 20 L, 230 VOverhead stirrer, HS-100 D unit, 20 L, 230 V8 mm180 mm90 mm145 mm200 - 3000 min-1Log in for availability1 * 1 ST621,00 €
- WITGDH.WOS01026Overhead stirrer, HS-100 D set, 20 L, 230 VOverhead stirrer, HS-100 D set, 20 L, 230 V8 mm180 mm90 mm145 mm200 - 3000 min-1Log in for availability1 * 1 ST806,00 €