0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Buffers
Chromatography Buffers
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™
:
Honeywell Chemicals
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™
:
Honeywell Chemicals
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™
Product Family Options
Description
pH value
Pack type
Pk
33582-1L
Ammonia buffer
10
Plastic bottle
1 L
1 * 1 L
1 * 1 L - 54,10 €
54,10 €
-
+
1 * 1 L
54,10 €
-
+
33582-6X1L
Ammonia buffer
10
Poly bottle
6 L
1 * 6 L
1 * 6 L - 424,00 €
424,00 €
-
+
1 * 6 L
424,00 €
-
+
EDTA disodium salt concentrate for dilution to 0.01 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
#: 32068.603
: VWR Chemicals
pH Electrodes, SenTix®
#: 662-1315
: WTW
VWR®, Bottles, Wide Neck, with Screw Cap
#: 215-5632
: VWR Collection