Centrifuge tubes, disposable, plain, with screw thread finish, KIMBLE®
Borosilicate glass, 33 expansion, low extractable.
- With conical-shaped bottoms on the 5, 10, and 15 ml sizes
- Bullet-nosed shape on the 50 ml size
- Chemical and temperature resistance caps withstand centrifugation up to 2980×g
- Tubes are plastic shrink-wrapped in modular trays for cleanliness and safety
: Glass conforms USP type I and ASTM E438, Type I, Class A requirements.
: Closures to be ordered separately.
Frequently Bought Together